Assay Details
| Gene Symbol: scinla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178131.3 | NP_835232.2 | 3 - 4 | 417 | 98 | |
| GenBank mRNA | BC094991.1 | - | 3 - 4 | 432 | 98 | |
| BC125898.1 | - | 3 - 4 | 388 | 98 | ||
| BC134865.1 | - | 3 - 4 | 417 | 98 | ||
| BC155581.1 | - | 3 - 4 | 554 | 98 | ||
More Information
Set Membership: |
|

scinla
Genomic Map