Assay Details
| Gene Symbol: rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178305.1 | NP_840090.1 | 3 - 4 | 689 | 59 | |
| GenBank mRNA | AB070208.1 | - | 3 - 4 | 689 | 59 | |
| BC162972.1 | - | 3 - 4 | 761 | 59 | ||
| BC162998.1 | - | 3 - 4 | 761 | 59 | ||
More Information
Set Membership: |
|

rapsn
Genomic Map