Assay Details
| Gene Symbol: lnpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199480.1 | NP_955774.1 | 6 - 7 | 562 | 127 | |
| GenBank mRNA | AY423028.1 | - | 6 - 7 | 491 | 127 | |
| BC050510.1 | - | 6 - 7 | 562 | 127 | ||
More Information
Set Membership: |
|

lnpa
Genomic Map