Assay Details
| Gene Symbol: jpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199575.2 | NP_955869.2 | 4 - 5 | 526 | 83 | |
| GenBank mRNA | AY394952.1 | - | 4 - 5 | 83 | ||
| BC045436.1 | - | 4 - 5 | 83 | |||
| BC066731.1 | - | 4 - 5 | 428 | 83 | ||
| BC154304.1 | - | 4 - 5 | 83 | |||
| BC155180.1 | - | 4 - 5 | 83 | |||
More Information
Set Membership: |
|

jpt2
Genomic Map