Assay Details
| Gene Symbol: tmem9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328536.1 | NP_001315465.1 | 5 - 6 | 468 | 84 | |
| NM_199866.2 | NP_956160.2 | 4 - 5 | 730 | 84 | ||
| XM_005167008.4 | XP_005167065.1 | 5 - 6 | 510 | 84 | ||
| XM_009303986.3 | XP_009302261.1 | 5 - 6 | 506 | 84 | ||
| GenBank mRNA | BC045338.1 | - | 4 - 5 | 676 | 84 | |
| BC164975.1 | - | 4 - 5 | 458 | 84 | ||
| EH445611.1 | - | 5 - 6 | 489 | 84 | ||
More Information
Set Membership: |
|

tmem9
Genomic Map