Assay Details
| Gene Symbol: osbpl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199872.2 | NP_956166.1 | 6 - 7 | 680 | 73 | |
| XM_005161947.3 | XP_005162004.1 | 6 - 7 | 689 | 73 | ||
| XM_005161948.4 | XP_005162005.1 | 6 - 7 | 641 | 73 | ||
| GenBank mRNA | AY394930.1 | - | 6 - 7 | 680 | 73 | |
| BC045502.1 | - | 6 - 7 | 681 | 73 | ||
More Information
Set Membership: |
|

osbpl2b
Genomic Map