Assay Details
| Gene Symbol: osbpl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199872.2 | NP_956166.1 | 9 - 10 | 1063 | 67 | |
| XM_005161947.3 | XP_005162004.1 | 9 - 10 | 1072 | 67 | ||
| XM_005161948.4 | XP_005162005.1 | 9 - 10 | 1024 | 67 | ||
| GenBank mRNA | AY394930.1 | - | 9 - 10 | 1063 | 67 | |
| BC045502.1 | - | 9 - 10 | 1064 | 67 | ||
More Information
Set Membership: |
|

osbpl2b
Genomic Map