Assay Details
| Gene Symbol: ppil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128330.1 | NP_001121802.1 | 8 - 9 | 832 | 66 | |
| XM_009294532.3 | XP_009292807.1 | 4 - 5 | 855 | 66 | ||
| XM_009294533.3 | XP_009292808.1 | 4 - 5 | 441 | 66 | ||
| GenBank mRNA | BC056814.1 | - | 8 - 9 | 814 | 66 | |
More Information
Set Membership: |
|

ppil4
Genomic Map