Assay Details
| Gene Symbol: ccdc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199985.2 | NP_956279.1 | 5 - 6 | 897 | 57 | |
| GenBank mRNA | BC049328.1 | - | 5 - 6 | 997 | 57 | |
| BC065606.1 | - | 5 - 6 | 897 | 57 | ||
| BC155816.1 | - | 5 - 6 | 955 | 57 | ||
More Information
Set Membership: |
|

ccdc6a
Genomic Map