Assay Details
| Gene Symbol: ccdc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199985.2 | NP_956279.1 | 7 - 8 | 1160 | 87 | |
| GenBank mRNA | BC049328.1 | - | 7 - 8 | 1260 | 87 | |
| BC065606.1 | - | 7 - 8 | 1160 | 87 | ||
| BC155816.1 | - | 7 - 8 | 1218 | 87 | ||
More Information
Set Membership: |
|

ccdc6a
Genomic Map