Assay Details
| Gene Symbol: krt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200080.2 | NP_956374.1 | 3 - 4 | 741 | 91 | |
| GenBank mRNA | AJ460000.1 | - | 3 - 4 | 604 | 91 | |
| AY423000.1 | - | 3 - 4 | 744 | 91 | ||
| BC067610.1 | - | 3 - 4 | 741 | 91 | ||
| BC154777.1 | - | 3 - 4 | 775 | 91 | ||
More Information
Set Membership: |
|

krt8
Genomic Map