Assay Details
| Gene Symbol: josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200151.2 | NP_956445.2 | 2 - 3 | 318 | 70 | |
| GenBank mRNA | BC044546.1 | - | 1 - 2 | 239 | 70 | |
| EH536544.1 | - | 2 - 3 | 348 | 70 | ||
More Information
Set Membership: |
|

josd2
Genomic Map