Assay Details
| Gene Symbol: desi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200208.1 | NP_956502.1 | 1 - 2 | 367 | 109 | |
| GenBank mRNA | BC045987.1 | - | 1 - 2 | 367 | 109 | |
| BC171711.1 | - | 1 - 2 | 318 | 109 | ||
More Information
Set Membership: |
|

desi1b
Genomic Map