Assay Details
| Gene Symbol: desi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200208.1 | NP_956502.1 | 5 - 6 | 691 | 86 | |
| GenBank mRNA | BC045987.1 | - | 5 - 6 | 691 | 86 | |
| BC171711.1 | - | 5 - 6 | 642 | 86 | ||
More Information
Set Membership: |
|

desi1b
Genomic Map