Assay Details
| Gene Symbol: otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200225.1 | NP_956519.1 | 3 - 4 | 479 | 71 | |
| GenBank mRNA | BC046060.1 | - | 3 - 4 | 479 | 71 | |
More Information
Set Membership: |
|

otud6b
Genomic Map