Assay Details
| Gene Symbol: usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200296.1 | NP_956590.1 | 6 - 7 | 624 | 96 | |
| XM_005157507.4 | XP_005157564.1 | 6 - 7 | 96 | |||
| XM_005157508.4 | XP_005157565.1 | 6 - 7 | 96 | |||
| XM_005157509.4 | XP_005157566.1 | 6 - 7 | 96 | |||
| XM_005157510.4 | XP_005157567.1 | 6 - 7 | 96 | |||
| XM_009291624.3 | XP_009289899.1 | 6 - 7 | 96 | |||
| XM_009291625.3 | XP_009289900.1 | 6 - 7 | 96 | |||
| XM_009291626.3 | XP_009289901.1 | 6 - 7 | 96 | |||
| XM_021481355.1 | XP_021337030.1 | 6 - 7 | 96 | |||
| XM_021481356.1 | XP_021337031.1 | 6 - 7 | 96 | |||
| XM_021481357.1 | XP_021337032.1 | 6 - 7 | 96 | |||
| GenBank mRNA | BC049481.1 | - | 6 - 7 | 624 | 96 | |
More Information
Set Membership: |
|

usf1
Genomic Map