Assay Details
| Gene Symbol: sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200372.1 | NP_956666.1 | 3 - 4 | 297 | 62 | |
| GenBank mRNA | BC053226.1 | - | 3 - 4 | 297 | 62 | |
| BC152223.1 | - | 4 - 5 | 320 | 62 | ||
More Information
Set Membership: |
|

sft2d1
Genomic Map