Assay Details
| Gene Symbol: mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200686.1 | NP_956980.1 | 2 - 3 | 566 | 60 | |
| GenBank mRNA | BC059182.1 | - | 2 - 3 | 566 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtfp1| Gene Symbol: mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200686.1 | NP_956980.1 | 2 - 3 | 566 | 60 | |
| GenBank mRNA | BC059182.1 | - | 2 - 3 | 566 | 60 | |
Set Membership: |
|