Assay Details
| Gene Symbol: pde6ha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305554.1 | NP_001292483.1 | 2 - 3 | 221 | 68 | |
| NM_200785.1 | NP_957079.1 | 2 - 3 | 204 | 68 | ||
| GenBank mRNA | BC059633.1 | - | 2 - 3 | 204 | 68 | |
| EH437109.1 | - | 2 - 3 | 233 | 68 | ||
| Gene Symbol: LOC110437775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021476972.1 | XP_021332647.1 | 2 - 3 | 227 | 68 | |
| XM_021476973.1 | XP_021332648.1 | 2 - 3 | 243 | 68 | ||
| GenBank mRNA | BC154316.1 | - | 2 - 3 | 207 | 68 | |
| BC163030.1 | - | 1 - 2 | 375 | 68 | ||
| BC163040.1 | - | 2 - 3 | 263 | 68 | ||
| EH433460.1 | - | 2 - 3 | 228 | 68 | ||
| EH459463.1 | - | 1 - 2 | 401 | 68 | ||
More Information
Set Membership: |
|

pde6ha
Genomic Map