Assay Details
| Gene Symbol: prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200805.1 | NP_957099.1 | 3 - 4 | 431 | 59 | |
| GenBank mRNA | BC059671.1 | - | 3 - 4 | 431 | 59 | |
| BC153616.1 | - | 3 - 4 | 434 | 59 | ||
More Information
Set Membership: |
|

prdx6
Genomic Map