Assay Details
| Gene Symbol: arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200818.1 | NP_957112.1 | 4 - 5 | 471 | 64 | |
| XM_005173309.3 | XP_005173366.1 | 4 - 5 | 543 | 64 | ||
| XM_021481069.1 | XP_021336744.1 | 4 - 5 | 543 | 64 | ||
| GenBank mRNA | BC059702.1 | - | 4 - 5 | 471 | 64 | |
More Information
Set Membership: |
|

arl13a
Genomic Map