Assay Details
| Gene Symbol: cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113609.1 | NP_001107081.1 | 6 - 7 | 407 | 110 | |
| XM_005166882.4 | XP_005166939.1 | 7 - 8 | 505 | 110 | ||
| GenBank mRNA | GU977276.1 | - | 7 - 8 | 492 | 110 | |
More Information
Set Membership: |
|

cenph
Genomic Map