Assay Details
| Gene Symbol: zeb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114551.2 | NP_001108023.2 | 4 - 5 | 546 | 142 | |
| NM_001135104.2 | NP_001128576.2 | 4 - 5 | 546 | 142 | ||
| XM_005167523.4 | XP_005167580.4 | 3 - 4 | 695 | 142 | ||
| XM_021478582.1 | XP_021334257.1 | 4 - 5 | 843 | 142 | ||
| XM_021478583.1 | XP_021334258.1 | 3 - 4 | 789 | 142 | ||
| GenBank mRNA | BC168514.1 | - | 4 - 5 | 526 | 142 | |
| EU379558.1 | - | 3 - 4 | 357 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021478583.1 transcripts.
Set Membership: |
|

zeb2a
Genomic Map