Assay Details
| Gene Symbol: zeb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114551.2 | NP_001108023.2 | 7 - 8 | 1069 | 90 | |
| NM_001135104.2 | NP_001128576.2 | 7 - 8 | 1069 | 90 | ||
| XM_005167523.4 | XP_005167580.4 | 6 - 7 | 1218 | 90 | ||
| XM_021478582.1 | XP_021334257.1 | 7 - 8 | 1366 | 90 | ||
| XM_021478583.1 | XP_021334258.1 | 6 - 7 | 1312 | 90 | ||
| GenBank mRNA | BC168514.1 | - | 7 - 8 | 1049 | 90 | |
| EU379558.1 | - | 6 - 7 | 880 | 90 | ||
More Information
Set Membership: |
|

zeb2a
Genomic Map