Assay Details
| Gene Symbol: serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114559.1 | NP_001108031.1 | 6 - 7 | 1073 | 84 | |
| GenBank mRNA | BC154797.1 | - | 6 - 7 | 1073 | 84 | |
| JQ801452.1 | - | 6 - 7 | 994 | 84 | ||
More Information
Set Membership: |
|

serpine1
Genomic Map