Assay Details
| Gene Symbol: cln8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114588.2 | NP_001108060.2 | 2 - 3 | 388 | 84 | |
| GenBank mRNA | BC154584.1 | - | 1 - 2 | 214 | 84 | |
More Information
Set Membership: |
|

cln8
Genomic Map