Assay Details
| Gene Symbol: coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114720.1 | NP_001108192.1 | 2 - 3 | 227 | 71 | |
| XM_005165486.4 | XP_005165543.1 | 2 - 3 | 387 | 71 | ||
| XM_005165487.4 | XP_005165544.1 | 2 - 3 | 245 | 71 | ||
| XM_017356012.2 | XP_017211501.1 | 2 - 3 | 245 | 71 | ||
| GenBank mRNA | BC155557.1 | - | 2 - 3 | 227 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005165486.4 transcripts.
Set Membership: |
|

coq4
Genomic Map