Assay Details
| Gene Symbol: insl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115053.2 | NP_001108525.2 | 1 - 2 | 280 | 76 | |
| GenBank mRNA | AM161139.1 | - | 1 - 2 | 280 | 76 | |
| AY834225.1 | - | 1 - 2 | 187 | 76 | ||
| EF685704.1 | - | 1 - 2 | 187 | 76 | ||
More Information
Set Membership: |
|

insl3
Genomic Map