Assay Details
| Gene Symbol: esama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115066.1 | NP_001108538.1 | 5 - 6 | 767 | 67 | |
| GenBank mRNA | CU638747.1 | - | 5 - 6 | 767 | 67 | |
| FJ205704.1 | - | 5 - 6 | 744 | 67 | ||
More Information
Set Membership: |
|

esama
Genomic Map