Assay Details
| Gene Symbol: gpa33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017353526.2 | XP_017209015.1 | 1 - 2 | 239 | 92 | |
| GenBank mRNA | BC117637.1 | - | 1 - 2 | 43 | 92 | |
| BC146710.1 | - | 1 - 2 | 67 | 92 | ||
| CU458914.1 | - | 1 - 2 | 66 | 92 | ||
More Information
Set Membership: |
|

gpa33a
Genomic Map