Assay Details
| Gene Symbol: ptbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122654.1 | NP_001116126.1 | 13 - 14 | 1366 | 82 | |
| XM_005155938.4 | XP_005155995.1 | 12 - 13 | 1295 | 82 | ||
| XM_009305880.2 | XP_009304155.1 | 13 - 14 | 1315 | 82 | ||
| GenBank mRNA | BC091854.1 | - | 13 - 14 | 1328 | 82 | |
More Information
Set Membership: |
|

ptbp1b
Genomic Map