Assay Details
| Gene Symbol: thsd7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122760.1 | NP_001116232.1 | 1 - 2 | 513 | 68 | |
| XM_005159680.4 | XP_005159737.1 | 1 - 2 | 507 | 68 | ||
| XM_009294351.3 | XP_009292626.1 | 1 - 2 | 507 | 68 | ||
| GenBank mRNA | BC151897.1 | - | 1 - 2 | 512 | 68 | |
More Information
Set Membership: |
|

thsd7aa
Genomic Map