Assay Details
| Gene Symbol: thsd7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122760.1 | NP_001116232.1 | 23 - 24 | 4947 | 55 | |
| XM_005159680.4 | XP_005159737.1 | 24 - 25 | 4947 | 55 | ||
| XM_009294351.3 | XP_009292626.1 | 24 - 25 | 4947 | 55 | ||
| GenBank mRNA | HQ267705.1 | - | 23 - 24 | 4431 | 55 | |
More Information
Set Membership: |
|

thsd7aa
Genomic Map