Assay Details
| Gene Symbol: thsd7aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122760.1 | NP_001116232.1 | 2 - 3 | 1465 | 72 | |
| XM_005159680.4 | XP_005159737.1 | 2 - 3 | 1459 | 72 | ||
| XM_009294351.3 | XP_009292626.1 | 2 - 3 | 1459 | 72 | ||
| GenBank mRNA | HQ267705.1 | - | 1 - 2 | 943 | 72 | |
More Information
Set Membership: |
|

thsd7aa
Genomic Map