Assay Details
| Gene Symbol: ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320519.1 | NP_001307448.1 | 2 - 3 | 213 | 57 | |
| XM_017354980.2 | XP_017210469.1 | 2 - 3 | 256 | 57 | ||
| XM_017354981.2 | XP_017210470.1 | 2 - 3 | 336 | 57 | ||
| GenBank mRNA | BC053185.1 | - | 2 - 3 | 446 | 57 | |
| BC165151.1 | - | 1 - 2 | 144 | 57 | ||
More Information
Set Membership: |
|

ttll1
Genomic Map