Assay Details
| Gene Symbol: rorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201067.1 | NP_957361.1 | 2 - 3 | 194 | 137 | |
| GenBank mRNA | BC051158.1 | - | 2 - 3 | 194 | 137 | |
| BC159203.1 | - | 2 - 3 | 137 | |||
More Information
Set Membership: |
|

rorab
Genomic Map