Assay Details
| Gene Symbol: rorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201067.1 | NP_957361.1 | 4 - 5 | 757 | 65 | |
| XM_009303216.3 | XP_009301491.1 | 5 - 6 | 1062 | 65 | ||
| XM_017356860.2 | XP_017212349.1 | 5 - 6 | 1067 | 65 | ||
| XM_021477531.1 | XP_021333206.1 | 4 - 5 | 1032 | 65 | ||
| GenBank mRNA | BC051158.1 | - | 4 - 5 | 757 | 65 | |
| BC159203.1 | - | 4 - 5 | 739 | 65 | ||
More Information
Set Membership: |
|

rorab
Genomic Map