Assay Details
| Gene Symbol: homer3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201113.1 | NP_957407.1 | 2 - 3 | 468 | 110 | |
| GenBank mRNA | BC045383.1 | - | 2 - 3 | 468 | 110 | |
| BC154233.1 | - | 439 | 110 | |||
| BC164401.1 | - | 1 - 2 | 190 | 110 | ||
More Information
Set Membership: |
|

homer3b
Genomic Map