Assay Details
| Gene Symbol: ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205558.1 | NP_991121.1 | 2 - 3 | 229 | 68 | |
| GenBank mRNA | BC056547.1 | - | 2 - 3 | 229 | 68 | |
| BC065471.1 | - | 2 - 3 | 229 | 68 | ||
| BC155185.1 | - | 2 - 3 | 240 | 68 | ||
More Information
Set Membership: |
|

ap1s2
Genomic Map