Assay Details
| Gene Symbol: pla1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207056.1 | NP_996939.1 | 4 - 5 | 806 | 60 | |
| GenBank mRNA | BC066406.1 | - | 4 - 5 | 806 | 60 | |
| BC155819.1 | - | 4 - 5 | 834 | 60 | ||
More Information
Set Membership: |
|

pla1a
Genomic Map