Assay Details
| Gene Symbol: camlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207090.2 | NP_996973.2 | 1 - 2 | 240 | 134 | |
| XM_021468793.1 | XP_021324468.1 | 1 - 2 | 243 | 128 | ||
| GenBank mRNA | BC066383.1 | - | 236 | 131 | ||
More Information
Set Membership: |
|

camlg
Genomic Map