Assay Details
| Gene Symbol: hey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212561.1 | NP_997726.1 | 2 - 3 | 203 | 57 | |
| GenBank mRNA | AJ510221.1 | - | 2 - 3 | 203 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hey1| Gene Symbol: hey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212561.1 | NP_997726.1 | 2 - 3 | 203 | 57 | |
| GenBank mRNA | AJ510221.1 | - | 2 - 3 | 203 | 57 | |
Set Membership: |
|