Assay Details
| Gene Symbol: poldip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212714.1 | NP_997879.1 | 7 - 8 | 988 | 64 | |
| XM_005157562.4 | XP_005157619.1 | 8 - 9 | 886 | 64 | ||
| GenBank mRNA | AY398395.1 | - | 7 - 8 | 988 | 64 | |
| BC045382.1 | - | 7 - 8 | 964 | 64 | ||
| BC152677.1 | - | 7 - 8 | 1009 | 64 | ||
| BC164775.1 | - | 7 - 8 | 813 | 64 | ||
More Information
Set Membership: |
|

poldip2
Genomic Map