Assay Details
| Gene Symbol: mpeg1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212737.1 | NP_997902.1 | 1 - 2 | 487 | 95 | |
| GenBank mRNA | BC055644.1 | - | 1 - 2 | 487 | 95 | |
More Information
Set Membership: |
|

mpeg1.1
Genomic Map