Assay Details
| Gene Symbol: otulina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213142.1 | NP_998307.1 | 1 - 2 | 234 | 103 | |
| GenBank mRNA | BC053249.1 | - | 1 - 2 | 234 | 103 | |
More Information
Set Membership: |
|

otulina
Genomic Map