Assay Details
| Gene Symbol: otulina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213142.1 | NP_998307.1 | 2 - 3 | 358 | 100 | |
| GenBank mRNA | BC053249.1 | - | 2 - 3 | 358 | 100 | |
More Information
Set Membership: |
|

otulina
Genomic Map