Assay Details
| Gene Symbol: gdpd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213201.1 | NP_998366.1 | 9 - 10 | 888 | 118 | |
| GenBank mRNA | BC064306.1 | - | 9 - 10 | 888 | 118 | |
More Information
Set Membership: |
|

gdpd3a
Genomic Map