Assay Details
| Gene Symbol: rab41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213365.1 | NP_998530.1 | 1 - 2 | 147 | 152 | |
| XM_005173245.4 | XP_005173302.1 | 1 - 2 | 197 | 152 | ||
| XM_005173246.4 | XP_005173303.1 | 1 - 2 | 202 | 152 | ||
| XM_005173247.4 | XP_005173304.1 | 1 - 2 | 199 | 152 | ||
| GenBank mRNA | BC058059.1 | - | 1 - 2 | 147 | 152 | |
| BC097051.1 | - | 1 - 2 | 174 | 152 | ||
| BC154221.1 | - | 1 - 2 | 92 | 152 | ||
More Information
Set Membership: |
|

rab41
Genomic Map