Assay Details
| Gene Symbol: elovl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328594.1 | NP_001315523.1 | 7 - 8 | 764 | 113 | |
| NM_213416.2 | NP_998581.2 | 5 - 6 | 657 | 113 | ||
| XM_009305889.3 | XP_009304164.1 | 8 - 9 | 774 | 113 | ||
| XM_009305890.3 | XP_009304165.1 | 5 - 6 | 547 | 113 | ||
| GenBank mRNA | BC049330.1 | - | 5 - 6 | 633 | 113 | |
| BC155205.1 | - | 4 - 5 | 407 | 113 | ||
| EH505140.1 | - | 7 - 8 | 717 | 113 | ||
| EH585991.1 | - | 5 - 6 | 632 | 114 | ||
More Information
Set Membership: |
|

elovl1b
Genomic Map