Assay Details
| Gene Symbol: elovl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328594.1 | NP_001315523.1 | 9 - 10 | 1017 | 70 | |
| NM_213416.2 | NP_998581.2 | 7 - 8 | 910 | 70 | ||
| XM_009305889.3 | XP_009304164.1 | 10 - 11 | 1027 | 70 | ||
| XM_009305890.3 | XP_009304165.1 | 7 - 8 | 800 | 70 | ||
| GenBank mRNA | BC049330.1 | - | 7 - 8 | 886 | 70 | |
| BC155205.1 | - | 6 - 7 | 660 | 70 | ||
More Information
Set Membership: |
|

elovl1b
Genomic Map